Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Diiodo-3-methyl-1H-pyrazole 98%

Supplier: AMBEED, INC
Warning

A123971-1G A123971-250MG A123971-25G A123971-5G
77161-488EA 31 USD InStock
77161-488 77161-490 77161-492 77161-494
4,5-Diiodo-3-methyl-1H-pyrazole 98%
4,5-Diiodo-3-methyl-1H-pyrazole
Precaución: for Research Use Only


Formula: C₄H₄I₂N₂
MDL Number: MFCD20484310
CAS Number: 6715-87-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty