Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3H-Imidazo[4,5-c]pyridin-2-ol 97%

Supplier: AMBEED, INC
Warning

A124677-10G A124677-1G A124677-5G
77189-834EA 128 USD InStock
77189-834 77189-836 77189-838
3H-Imidazo[4,5-c]pyridin-2-ol 97%
3H-Imidazo[4,5-c]pyridin-2-ol
Precaución: for Research Use Only


Formula: C₆H₅N₃O
MDL Number: MFCD18872694
CAS Number: 7397-68-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty