Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

LDN193189 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A125436-10MG A125436-100MG A125436-250MG A125436-50MG A125436-5MG
77161-590EA 59 USD InStock
77161-590 77611-068 77683-115 77683-116 77683-117
LDN193189 98%, Ambeed.Inc
LDN193189
Precaución: For research use only.


Formula: C₂₅H₂₂N₆
MDL Number: MFCD17392570
CAS Number: 1062368-24-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty