Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxynicotinic acid 98%

Supplier: AMBEED, INC
Warning

A126843-1000G A126843-100G A126843-25G A126843-500G A126843-5G
77189-910EA 211.01 USD InStock
77189-910 77189-912 77189-914 77189-916 77189-918
2-Methoxynicotinic acid 98%
2-Methoxynicotinic acid
Precaución: for Research Use Only


Formula: C₇H₇NO₃
MW: 153.14
MDL Number: MFCD00085110
CAS Number: 16498-81-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty