Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tiopronin 98%, Ambeed.Inc

Supplier: AMBEED, INC
Danger

A127313-100G A127313-10G A127313-1G A127313-5G A127313-25G
77161-654EA 132 USD InStock
77161-654 77161-656 77161-658 77161-660 77683-129
Tiopronin 98%, Ambeed.Inc
Tiopronin
Precaución: For research use only.


Formula: C₅H₉NO₃S
MW: 163.19 g/mol
MDL Number: MFCD00004861
CAS Number: 1953-02-2
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty