Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nisoldipine 97%

Supplier: AMBEED, INC
Warning

A127509-100G A127509-100MG A127509-1G A127509-1MG A127509-250MG A127509-25G A127509-5G
77189-946EA 487.01 USD InStock
77189-946 77189-948 77189-950 77189-952 77189-954 77189-956 77189-958
Nisoldipine 97%
Nisoldipine
Precaución: for Research Use Only


Formula: C₂₀H₂₄N₂O₆
MW: 388.42 g/mol
MDL Number: MFCD00478055
CAS Number: 63675-72-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty