Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyldimethyltetradecylammonium chloride dihydrate 98%

Supplier: AMBEED, INC
Danger

A128053-1G A128053-25G A128053-5G
77161-718EA 5 USD InStock
77161-718 77161-720 77161-722
Benzyldimethyltetradecylammonium chloride dihydrate 98%
Benzyldimethyltetradecylammonium chloride dihydrate
Precaución: for Research Use Only


Formula: C₂₃H₄₂ClN·2H₂O
MW: 404.0699 g/mol
MDL Number: MFCD00149985
CAS Number: 147228-81-7
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty