Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N'-Di-Boc-guanidine 95%

Supplier: AMBEED, INC
Warning

A130740-10G A130740-1G A130740-250MG A130740-25G A130740-5G
77190-048EA 67 USD InStock
77190-048 77190-050 77190-052 77190-054 77190-056
N,N'-Di-Boc-guanidine 95%
N,N'-Di-Boc-guanidine
Precaución: for Research Use Only


Formula: C₁₁H₂₁N₃O₄
MW: 259.30 g/mol
MDL Number: MFCD01863604
CAS Number: 154476-57-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty