Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-2-piperidineacetic acid 95%

Supplier: AMBEED, INC
Warning

A131697-1G A131697-250MG A131697-5G
77190-068EA 14 USD InStock
77190-068 77190-070 77190-072
1-Boc-2-piperidineacetic acid 95%
1-Boc-2-piperidineacetic acid
Precaución: for Research Use Only


Formula: C₁₂H₂₁NO₄
MDL Number: MFCD02179000
CAS Number: 149518-50-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty