Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methylnicotinamide 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A131881-10G A131881-1G A131881-5G A131881-25G
77161-918EA 25 USD InStock
77161-918 77161-920 77161-922 77683-169
6-Methylnicotinamide 98%, Ambeed.Inc
6-Methylnicotinamide
Precaución: For research use only.


Formula: C₇H₈N₂O
MDL Number: MFCD00006342
CAS Number: 6960-22-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty