Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-(Phenylamino)phenyl)ethanone 98%

Supplier: AMBEED, INC
Warning

A132106-100MG A132106-1G A132106-250MG A132106-5G
77190-092EA 22 USD InStock
77190-092 77190-094 77190-096 77745-945
1-(4-(Phenylamino)phenyl)ethanone 98%
1-(4-(Phenylamino)phenyl)ethanone
Precaución: for Research Use Only


Formula: C₁₄H₁₃NO
MW: 211.26 g/mol
MDL Number: MFCD09032841
CAS Number: 23600-83-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty