Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Pyrimidinecarboxylic acid 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A132586-10G A132586-1G A132586-25G A132586-5G A132586-250MG
77161-934EA 23 USD InStock
77161-934 77161-936 77161-938 77161-940 77683-176
2-Pyrimidinecarboxylic acid 97%, Ambeed.Inc
2-Pyrimidinecarboxylic acid
Precaución: For research use only.


Formula: C₅H₄N₂O₂
MDL Number: MFCD00856161
CAS Number: 31519-62-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty