Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methyl-2-indoleboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A134725-1G A134725-250MG A134725-5G
77162-042EA 125 USD InStock
77162-042 77162-044 77162-046
6-Methyl-2-indoleboronic acid pinacol ester 98%
6-Methyl-2-indoleboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₅H₂₀BNO₂
MW: 257.1358 g/mol
MDL Number: MFCD11858378
CAS Number: 912332-45-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty