Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ARN-509 98%

Supplier: AMBEED, INC
Danger

A136293-100MG A136293-10MG A136293-1G A136293-250MG A136293-5G A136293-5MG
77162-102EA 21 USD InStock
77162-102 77162-104 77162-106 77162-108 77162-110 77162-112
ARN-509 98%
ARN-509
Precaución: for Research Use Only


Formula: C₂₁H₁₅F₄N₅O₂S
MW: 477.44 g/mol
MDL Number: MFCD22380626
CAS Number: 956104-40-8
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty