Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluorophenylhydrazine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A137065-100G A137065-1G A137065-25G A137065-5G
77162-148EA 127 USD InStock
77162-148 77162-150 77162-152 77162-154
4-Bromo-2-fluorophenylhydrazine hydrochloride 98%
4-Bromo-2-fluorophenylhydrazine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₆BrFN₂·ClH
MDL Number: MFCD03094166
CAS Number: 502496-24-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty