Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(((2-Ethylhexyl)oxy)carbonyl)benzoic acid 97%

Supplier: AMBEED, INC
Warning

A137885-10G A137885-1G A137885-5G
77162-194EA 697 USD InStock
77162-194 77162-196 77162-198
4-(((2-Ethylhexyl)oxy)carbonyl)benzoic acid 97%
4-(((2-Ethylhexyl)oxy)carbonyl)benzoic acid
Precaución: for Research Use Only


Formula: C₁₆H₂₂O₄
MDL Number: MFCD02752454
CAS Number: 155603-50-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty