Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Carboxymethylmorpholine-4-carboxylic acid tert-butyl ester 97%

Supplier: AMBEED, INC
Warning

A137937-100MG A137937-1G A137937-250MG
77162-200EA 54 USD InStock
77162-200 77162-202 77162-204
3-Carboxymethylmorpholine-4-carboxylic acid tert-butyl ester 97%
3-Carboxymethylmorpholine-4-carboxylic acid tert-butyl ester
Precaución: for Research Use Only


Formula: C₁₁H₁₉NO₅
MDL Number: MFCD04114891
CAS Number: 859155-89-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty