Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-1-naphthaleneboronic acid 97%

Supplier: AMBEED, INC
Warning

A138505-100MG A138505-10G A138505-1G A138505-250MG A138505-25G A138505-5G
77190-294EA 8 USD InStock
77190-294 77190-296 77190-298 77190-300 77190-302 77190-304
4-Chloro-1-naphthaleneboronic acid 97%
4-Chloro-1-naphthaleneboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₈BClO₂
MDL Number: MFCD18411295
CAS Number: 147102-97-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty