Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,5R)-5-(4-((2-Fluorobenzyl)oxy)phenyl)pyrrolidine-2-carboxamide 98+%

Supplier: AMBEED, INC
Warning

A138601-10MG A138601-1MG A138601-25MG A138601-5MG
77162-228EA 165 USD InStock
77162-228 77162-230 77162-232 77162-234
(2S,5R)-5-(4-((2-Fluorobenzyl)oxy)phenyl)pyrrolidine-2-carboxamide 98+%
(2S,5R)-5-(4-((2-Fluorobenzyl)oxy)phenyl)pyrrolidine-2-carboxamide
Precaución: for Research Use Only


Formula: C₁₈H₁₉FN₂O₂
MW: 314.35 g/mol
MDL Number: MFCD22580419
CAS Number: 934240-30-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty