Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Butoxy-4-iodobenzene 97%

Supplier: AMBEED, INC
Warning

A138889-1G A138889-250MG A138889-5G
77190-310EA 24 USD InStock
77190-310 77190-312 77190-314
1-Butoxy-4-iodobenzene 97%
1-Butoxy-4-iodobenzene
Precaución: for Research Use Only


Formula: C₁₀H₁₃IO
MDL Number: MFCD09878143
CAS Number: 96693-04-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty