Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Brimonidine tartrate 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A139688-100MG A139688-1G
77162-250EA 19 USD InStock
77162-250 77162-252 77162-254 77683-217
Brimonidine tartrate 98%, Ambeed.Inc
Brimonidine tartrate
Precaución: For research use only.


Formula: C₁₅H₁₆BrN₅O₆
MDL Number: MFCD07773072
CAS Number: 70359-46-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty