Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-6-aminopyridin-2-ylcarbamate 97%

Supplier: AMBEED, INC
Warning

A140404-1G A140404-250MG A140404-5G
77162-274EA 35 USD InStock
77162-274 77162-276 77162-278
tert-Butyl-6-aminopyridin-2-ylcarbamate 97%
tert-Butyl-6-aminopyridin-2-ylcarbamate
Precaución: for Research Use Only


Formula: C₁₀H₁₅N₃O₂
MW: 209.2500 g/mol
MDL Number: MFCD09037865
CAS Number: 322690-31-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty