Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Isobutyldihydro-2H-pyran-2,6(3H)-dione 95%

Supplier: AMBEED, INC
Warning

A141653-10G A141653-1G A141653-25G A141653-5G
77162-336EA 8 USD InStock
77162-336 77162-338 77162-340 77162-342
4-Isobutyldihydro-2H-pyran-2,6(3H)-dione 95%
4-Isobutyldihydro-2H-pyran-2,6(3H)-dione
Precaución: for Research Use Only


Formula: C₉H₁₄O₃
MDL Number: MFCD15529750
CAS Number: 185815-59-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty