Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Chlorobenzamido)-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid 98%

Supplier: AMBEED, INC
Warning

A141959-100G A141959-1G A141959-1MG A141959-25G A141959-5G
77162-358EA 150 USD InStock
77162-358 77162-360 77162-362 77162-364 77162-366
2-(4-Chlorobenzamido)-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid 98%
2-(4-Chlorobenzamido)-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid
Precaución: for Research Use Only


Formula: C₁₉H₁₅ClN₂O₄
MDL Number: MFCD00866895
CAS Number: 90098-04-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty