Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-3-Aminocyclopentanecarboxylic acid hydrochloride 95%

Supplier: AMBEED, INC
Warning

A142986-1G A142986-250MG
77162-426EA 32 USD InStock
77162-426 77162-428
cis-3-Aminocyclopentanecarboxylic acid hydrochloride 95%
cis-3-Aminocyclopentanecarboxylic acid hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₂ClNO₂
MDL Number: MFCD23106173
CAS Number: 19042-35-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty