Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Triphenylphosphoranylidene)acetonitrile 97%

Supplier: AMBEED, INC
Danger

A144437-10G A144437-1G A144437-25G A144437-5G
77190-554EA 37 USD InStock
77190-554 77190-556 77190-558 77190-560
(Triphenylphosphoranylidene)acetonitrile 97%
(Triphenylphosphoranylidene)acetonitrile
Precaución: for Research Use Only


Formula: C₂₀H₁₆NP
MDL Number: MFCD00567633
CAS Number: 16640-68-9
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty