Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-nitrobenzenemethanol 98%

Supplier: AMBEED, INC
Warning

A145094-100MG A145094-1G A145094-250MG A145094-5G
77190-574EA 6 USD InStock
77190-574 77190-576 77190-578 77746-015
2-Amino-4-nitrobenzenemethanol 98%
2-Amino-4-nitrobenzenemethanol
Precaución: for Research Use Only


Formula: C₇H₈N₂O₃
MDL Number: MFCD01664410
CAS Number: 78468-34-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty