Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,4S)-5-([1,1'-Biphenyl]-4-yl)-4-(3-carboxypropanamido)-2-methylpentanoic acid 97%

Supplier: AMBEED, INC
Danger

A145106-10MG A145106-1MG A145106-25MG A145106-50MG A145106-5MG
77190-580EA 57 USD InStock
77190-580 77190-582 77190-584 77190-586 77190-588
(2R,4S)-5-([1,1'-Biphenyl]-4-yl)-4-(3-carboxypropanamido)-2-methylpentanoic acid 97%
(2R,4S)-5-([1,1'-Biphenyl]-4-yl)-4-(3-carboxypropanamido)-2-methylpentanoic acid
Precaución: for Research Use Only


Formula: C₂₂H₂₅NO₅
MDL Number: MFCD00921225
CAS Number: 149709-44-4
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty