Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-methylbenzofuro[3,2-d]pyrimidine 97%

Supplier: AMBEED, INC
Warning

A145476-100MG A145476-250MG
77190-598EA 7 USD InStock
77190-598 77190-600
4-Chloro-2-methylbenzofuro[3,2-d]pyrimidine 97%
4-Chloro-2-methylbenzofuro[3,2-d]pyrimidine
Precaución: For research use only.


Formula: C₁₁H₇ClN₂O
MDL Number: MFCD01202941
CAS Number: 39786-40-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty