Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 2-((2-amino-6-oxo-1H-purin-9(6H)-yl)methoxy)ethanolate dihydrate 95%

Supplier: AMBEED, INC
Warning

A147424-10G A147424-1G
77162-604EA 38 USD InStock
77162-604 77162-606 77162-608
Sodium 2-((2-amino-6-oxo-1H-purin-9(6H)-yl)methoxy)ethanolate dihydrate 95%
Sodium 2-((2-amino-6-oxo-1H-purin-9(6H)-yl)methoxy)ethanolate dihydrate
Precaución: for Research Use Only


Formula: C₈H₁₄N₅NaO₅
MW: 283.21 g/mol
MDL Number: MFCD28902495
CAS Number: 261506-45-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty