Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Difluorobenzamide 97%

Supplier: AMBEED, INC
Warning

A147994-10G A147994-25G
77162-628EA 17 USD InStock
77162-628 77162-630 77162-632
2,5-Difluorobenzamide 97%
2,5-Difluorobenzamide
Precaución: for Research Use Only


Formula: C₇H₅F₂NO
MW: 157.12 g/mol
MDL Number: MFCD00015548
CAS Number: 85118-03-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty