Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-acetyl-3-(dimethylamino)acrylate 97%

Supplier: AMBEED, INC
Warning

A148937-10G A148937-1G A148937-250MG A148937-5G
77162-670EA 118 USD InStock
77162-670 77162-672 77162-674 77162-676
Methyl-2-acetyl-3-(dimethylamino)acrylate 97%
Methyl-2-acetyl-3-(dimethylamino)acrylate
Precaución: for Research Use Only


Formula: C₈H₁₃NO₃
MDL Number: MFCD08689663
CAS Number: 203186-56-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty