Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N1,N3-Bis(2,2,6,6-tetramethylpiperidin-4-yl)isophthalamide 99+%

Supplier: AMBEED, INC
Warning

A149067-10G A149067-1G A149067-250MG A149067-25G A149067-5G
77190-710EA 52 USD InStock
77190-710 77190-712 77190-714 77190-716 77190-718
N1,N3-Bis(2,2,6,6-tetramethylpiperidin-4-yl)isophthalamide 99+%
N1,N3-Bis(2,2,6,6-tetramethylpiperidin-4-yl)isophthalamide
Precaución: for Research Use Only


Formula: C₂₆H₄₂N₄O₂
MDL Number: MFCD09751152
CAS Number: 42774-15-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty