Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Di(1H-imidazol-1-yl)butane 95%

Supplier: AMBEED, INC
Warning

A149858-100MG A149858-1G A149858-250MG A149858-5G
77162-720EA 11 USD InStock
77162-720 77162-722 77162-724 77162-726
1,4-Di(1H-imidazol-1-yl)butane 95%
1,4-Di(1H-imidazol-1-yl)butane
Precaución: for Research Use Only


Formula: C₁₀H₁₄N₄
MDL Number: MFCD00608317
CAS Number: 69506-86-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty