Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-Ethyl 4-(aminomethyl)cyclohexanecarboxylate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A150907-10G A150907-1G A150907-250MG
77162-792EA 99 USD InStock
77162-792 77162-794 77162-796
trans-Ethyl 4-(aminomethyl)cyclohexanecarboxylate hydrochloride 98%
trans-Ethyl 4-(aminomethyl)cyclohexanecarboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₂₀ClNO₂
MDL Number: MFCD28134591
CAS Number: 19878-18-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty