Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrazolo[1,5-a]pyridine 97%

Supplier: AMBEED, INC
Warning

A151078-100MG A151078-10G A151078-1G A151078-250MG A151078-25G A151078-5G
77162-806EA 10 USD InStock
77162-806 77162-808 77162-810 77162-812 77162-814 77162-816
Pyrazolo[1,5-a]pyridine 97%
Pyrazolo[1,5-a]pyridine
Precaución: for Research Use Only


Formula: C₇H₆N₂
MW: 118.14 g/mol
MDL Number: MFCD08752622
CAS Number: 274-56-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty