Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Methoxy-ethoxy)-phenol 97%

Supplier: AMBEED, INC
Warning

A154034-100MG A154034-1G A154034-250MG A154034-25G
77162-908EA 11 USD InStock
77162-908 77162-910 77162-912 77162-914
4-(2-Methoxy-ethoxy)-phenol 97%
4-(2-Methoxy-ethoxy)-phenol
Precaución: for Research Use Only


Formula: C₉H₁₂O₃
MDL Number: MFCD04114941
CAS Number: 51980-60-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty