Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A155693-100G A155693-25G A155693-5G
77190-910EA 22 USD InStock
77190-910 77190-912 77190-914
4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride 97%
4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₀ClNS
MDL Number: MFCD05861485
CAS Number: 28783-41-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty