Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AZD-9291 99%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A157172-10MG A157172-1G A157172-1MG A157172-250MG A157172-5G A157172-5MG A157172-25G
77163-026EA 9 USD InStock
77163-026 77163-028 77163-030 77163-032 77163-034 77163-036 77683-312
AZD-9291 99%, Ambeed.Inc
AZD-9291
Precaución: For research use only.


Formula: C₂₈H₃₃N₇O₂
MW: 499.61 g/mol
MDL Number: MFCD27988062
CAS Number: 1421373-65-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty