Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Diiodo-4-methylbenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A157263-100MG A157263-250MG
77163-044EA 11 USD InStock
77163-044 77163-046
3,5-Diiodo-4-methylbenzaldehyde 97%
3,5-Diiodo-4-methylbenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₆I₂O
MDL Number: MFCD09032526
CAS Number: 861118-00-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty