Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methylamino)propanoic acid hydrochloride 98%

Supplier: AMBEED, INC
Warning

A157320-1G A157320-250MG A157320-5G
77163-056EA 40 USD InStock
77163-056 77163-058 77163-060
3-(Methylamino)propanoic acid hydrochloride 98%
3-(Methylamino)propanoic acid hydrochloride
Precaución: for Research Use Only


Formula: C₄H₁₀ClNO₂
MW: 139.58 g/mol
MDL Number: MFCD03490507
CAS Number: 65845-56-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty