Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dibromo-1,4-difluorobenzene 95%

Supplier: AMBEED, INC
Warning

A159481-100MG A159481-10G A159481-1G A159481-250MG A159481-25G A159481-5G
77163-148EA 5 USD InStock
77163-148 77163-150 77163-152 77163-154 77163-156 77163-158
2,3-Dibromo-1,4-difluorobenzene 95%
2,3-Dibromo-1,4-difluorobenzene
Precaución: for Research Use Only


Formula: C₆H₂Br₂F₂
MW: 271.885 g/mol
MDL Number: MFCD12922620
CAS Number: 179737-33-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty