Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(1-Piperidinyl)benzeneboronic acid pinacol ester 96%

Supplier: AMBEED, INC
Warning

A160175-100MG A160175-250MG
77191-054EA 28 USD InStock
77191-054 77191-056
4-(1-Piperidinyl)benzeneboronic acid pinacol ester 96%
4-(1-Piperidinyl)benzeneboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₇H₂₆BNO₂
MW: 287.21 g/mol
MDL Number: MFCD07368524
CAS Number: 852227-96-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty