Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Acetylpyrene 98+%

Supplier: AMBEED, INC
Warning

A161782-10G A161782-1G A161782-5G
77191-104EA 220 USD InStock
77191-104 77191-106 77191-108
1-Acetylpyrene 98+%
1-Acetylpyrene
Precaución: for Research Use Only


Formula: C₁₈H₁₂O
MDL Number: MFCD00021663
CAS Number: 3264-21-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty