Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tubastatin A 98%

Supplier: AMBEED, INC
Warning

A161831-10MG A161831-1MG A161831-50MG A161831-5MG
77191-110EA 48 USD InStock
77191-110 77191-112 77191-114 77191-116
Tubastatin A 98%
Tubastatin A
Precaución: for Research Use Only


Formula: C₂₀H₂₁N₃O₂
MDL Number: MFCD18071463
CAS Number: 1252003-15-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty