Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzene-1,2,4,5-tetraol 95%

Supplier: AMBEED, INC
Warning

A162888-100MG A162888-1G A162888-250MG A162888-5G
77191-152EA 18 USD InStock
77191-152 77191-154 77191-156 77191-158
Benzene-1,2,4,5-tetraol 95%
Benzene-1,2,4,5-tetraol
Precaución: for Research Use Only


Formula: C₆H₆O₄
MDL Number: MFCD19441381
CAS Number: 636-32-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty