Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,6-Dibromohexane 97%

Supplier: AMBEED, INC
Danger

A163362-25G A163362-1000G A163362-500G
77191-184EA 12 USD InStock
77191-184 77567-384 77746-109
1,6-Dibromohexane 97%
1,6-Dibromohexane
Precaución: For research use only.


Formula: BrH₂C(CH₂)₄CH₂Br
MDL Number: MFCD00000272
CAS Number: 629-03-8
UN: 3082
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty