Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-(2-(4-Chlorophenyl)thiazol-4-yl)ethyl)butyramide 98%

Supplier: AMBEED, INC
Warning

A169042-10MG A169042-1MG A169042-50MG A169042-5MG
77163-446EA 21 USD InStock
77163-446 77163-448 77163-450 77163-452
N-(2-(2-(4-Chlorophenyl)thiazol-4-yl)ethyl)butyramide 98%
N-(2-(2-(4-Chlorophenyl)thiazol-4-yl)ethyl)butyramide
Precaución: for Research Use Only


Formula: C₁₅H₁₇ClN₂OS
MDL Number: MFCD08009374
CAS Number: 932986-18-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty