Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Pyridyl)ethylamine 97%

Supplier: AMBEED, INC
Danger

A169086-100MG A169086-10G A169086-1G A169086-250MG A169086-5G
77163-454EA 16 USD InStock
77163-454 77163-456 77163-458 77163-460 77163-462
1-(2-Pyridyl)ethylamine 97%
1-(2-Pyridyl)ethylamine
Precaución: for Research Use Only


Formula: C₇H₁₀N₂
MDL Number: MFCD05215238
CAS Number: 42088-91-5
UN: 2922
ADR: 8,6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty