Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

LGK974 98+%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A169157-10MG A169157-1MG A169157-250MG A169157-5MG A169157-50MG
77163-466EA 56 USD InStock
77163-466 77163-468 77163-470 77163-472 77683-386
LGK974 98+%, Ambeed.Inc
LGK974
Precaución: For research use only.


Formula: C₂₃H₂₀N₆O
MDL Number: MFCD27501026
CAS Number: 1243244-14-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty